Order Entry
United States
Orders LinkContactUsLinkComponent
chemagic Prepito

chemagic Prepito

chemagic Prepito automated instrument, Fuses on inlet: T 3.15 A, 250 V; Noise emission: <70 dB
Catalog # 77938-574
New Product
Supplier #:  2022-0030
chemagic Prepito
Catalog # 77938-574
New Product
chemagic Prepito
chemagic Prepito

chemagic Prepito automated instrument, Fuses on inlet: T 3.15 A, 250 V; Noise emission: <70 dB

Price
$31,109.15
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Benchtop instrument for automated nucleic acid purification from 1 to 12 samples. Based on chemagic separation technology that obtains high integrity, yields and purity of nucleic acids suitable for long read sequencing, NGS, PCR, genotyping arrays and more.

  • High integrity, yield, and purity of RNA and DNA

  • 1 to 12 samples per run, input volumes up to 1 ml
  • Small benchtop solution
  • Integrated buffer dispensing
  • Revolutionary resuspension technology that avoids shearing of nucleic acids
  • Standard plastic consumables (included in the Prepito kits)
  • Prepito kits and pre-installed protocols for a huge variety of sample materials
  • Barcode reading
  • LIMS-compatible log files
  • USB port

The chemagic™ Prepito™ (RUO) is for automated nucleic acid purification from diverse sample types. Used with chemagic Prepito kits, this compact benchtop solution obtains ready-to-use nucleic acids for challenging downstream assays. This cost-effective solution avoids the use of expensive cartridges and can process up to 12 samples per run with minimal hands-on intervention.

High integrity yields and purity of nucleic acids suitable for long read sequencing, MLPA, NGS, PCR, genotyping arrays and more.
Traceable sample preparation from barcode scanning and USB port for transfer of LIMS-compatible log files.
Automated buffer dispensing reduces hands-on time and avoids expensive cartridges.

 : For research use only. Not for use in diagnostic procedures.

Specifications
  • Description:
    chemagic Prepito automated instrument
  • Details:
    Fuses on inlet: T 3.15 A, 250 V; Noise emission: <70 dB
  • Cat. No.:
    77938-574
  • W×D×H:
    520×620×550 mm
  • Electrical:
    110 - 240 VAC, 50 - 60 Hz
  • Power:
    100 VA
  • Weight:
    45 kg