- Description:Wall mounted wipes and bottle holder
- W×D×H:16×4.5×12"
Palbam’s wipes and bottle holder is a stainless steel, wall-mountable solution designed for cleanrooms. They ensure organized, contamination-free storage of wipes and bottles, promoting efficiency, accessibility, and compliance in controlled environments.
Stainless steel 304 with electropolished finish
- Holds wipes and IPA bottles
The gownroom is often the first impression for employees and visitors alike. Palbam Class products are guaranteed to ensure that your company will give the best possible impression, as well as serving all ergonomic, cleanliness, and functional requirements.