ContactUsLinkComponent
Cleanroom Wipe Down Table
Catalog # 77898-322
New Product
undefined
Cleanroom Wipe Down Table
Catalog # 77898-322
Supplier Number:  WDT-0960
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Worksurface material:
    304 stainless steel
  • Description:
    Cleanroom wipe down table
  • W×D×H:
    36×24×36"
  • Worksurface color/finish:
    Electropolished
Specifications
About this item
Palbam’s cleanroom wipe down table is a stainless steel, ergonomic solution designed for cleanrooms.
  • Stainless steel 304 with electropolished finish
  • Includes hole in table top for waste disposal
  • Includes removable plastic bin
It provides a durable, contamination-free surface for efficient wipe-down procedures, ensuring compliance and cleanliness in controlled environments. The wipe down table supports the wipe-down procedure perfectly, featuring a hole in the tabletop for easy waste disposal. The cabinet beneath the table and waste disposal hole includes a removable plastic bin for convenient waste management.
The gownroom is often the first impression for employees and visitors alike, that they are entering into a special area. Palbam Class products are guaranteed to ensure that your company will give the best possible impression, as well as serving all the ergonomic, cleanliness and functional requirements.
Ordering information:
Available in several dimensions; contact your Avantor representative for more information.