ContactUsLinkComponent
Onset Air Velocity and Temperature Remote Head Sensor
Onset Air Velocity and Temperature Remote Head Sensor
New Product
Certificates
undefined
Onset Air Velocity and Temperature Remote Head Sensor
New Product
Specifications
  • Catalog No:
  • 470358-380
  • 470358-384
  • 470358-382
  • Temperature Range:
  • 0...60 °C (32...140 °F)
  • 0...60 °C (32...140 °F)
  • 0...60 °C (32...140 °F)
  • Response Time:
  • 400 ms
  • 400 ms
  • 400 ms