Specifications
- Subjects:Environmental
- Description:Aquatic detectives kit
- Grade:HS/College
Specifications
About this item
Explore the impact of water pollution on wildlife and human populations.
Study how factory discharge impacts bodies of water
Students test simulated water samples to determine what contaminants are present and which factories contribute.