Specifications
- Subjects:Biotech
- Description:Genetic finch diversity kit
- Grade:HS/College
Specifications
About this item
Investigate the genetic basis of beak size variation in Galapagos finches before and after a drought event using RFLP analysis.
Introduces PCR-RFLP analysis to analyze specific nucleotide sequences
- Explore how electrophoresis separates differently-sized molecules
Meets NGSS HS DCI LS1.A, LS3.A, and LS3.B standards.