Order Entry
United States
ContactUsLinkComponent
Bacteriophage Recombinant gp32 Glycerol-Free (from E. coli)
Bacteriophage Recombinant gp32 Glycerol-Free (from E. coli)
Certificates
undefined
Bacteriophage Recombinant gp32 Glycerol-Free (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Species:
    Bacteriophage
  • Storage Conditions:
    Store at –80 °C
  • Protein/Peptide Name:
    T4 gp32 protein, glycerol-free
  • Shipping Temperature:
    Dry Ice
  • Tested Applications:
    RPA
  • Protein/Peptide Type:
    Recombinant

Specifications

Frequently Bought Together