Order Entry
United States
ContactUsLinkComponent
 
Bacteriophage Recombinant UvsY Glycerol-Free (from E. coli)
 
undefined
Bacteriophage Recombinant UvsY Glycerol-Free (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Bacteriophage
  • Storage Conditions:
    Store at –80 °C
  • Protein Synonyms:
    T4 UvsY Protein, Glycerol-free
  • Protein/Peptide Name:
    UvsY
  • Tested Applications:
    RPA

 

Frequently Bought Together