Order Entry
United States
ContactUsLinkComponent
igScript™ Reverse Transcriptase, Glycerol-free, Intact Genomics, Inc.
igScript™ Reverse Transcriptase, Glycerol-free, Intact Genomics, Inc.
Supplier:  Intact Genomics
Certificates
undefined
igScript™ Reverse Transcriptase, Glycerol-free, Intact Genomics, Inc.
Supplier:  Intact Genomics

Specifications

  • Conjugation:
    Unconjugated
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Virus
  • Storage Conditions:
    Store at –80 °C
  • Enzyme Name:
    Reverse transcriptase
  • Enzyme Synonyms:
    Glycerol-free, RT
  • Tested Applications:
    RT-qPCR

Specifications

Frequently Bought Together