Order Entry
United States
ContactUsLinkComponent
PEmax Enzyme, Intact Genomics
PEmax Enzyme, Intact Genomics
Supplier:  Intact Genomics
Certificates
undefined
PEmax Enzyme, Intact Genomics
Supplier:  Intact Genomics

Specifications

  • Conjugation:
    Unconjugated
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Bacteria
  • Storage Conditions:
    Store at –20 °C
  • Enzyme Name:
    PEmax
  • Enzyme Synonyms:
    PEmax Enzyme
  • Tested Applications:
    CRISPR and gene editing

Specifications

Frequently Bought Together