xTIP™ Sterile Filter Tip Reload Rainin LTS Compatible Pipette Tips
Supplier: Biotix
New Product
Certificates
About this item
Biotix sterile xTIP™ filter tip reload rainin LTS compatible pipette tips with Flexfit™ to lower insertion and ejection force by 70%.
xTIP™ filter tip reloads are manufactured in sustainable benchtop and user friendly packaging
- Reduce plastic waste by 30% and easily refill racks
- Tips fit rainin LTS and xPipette pipettes
- Pure resin made of virgin polypropylene
- Certified free of RNase, DNase, pyrogens and PCR inhibitors
FlexFit™ technology features flexible proximal end to ensure secure seal with 66 - 80% less insertion force than competitor tips. Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip.
Ordering information:
Tip refills only. Empty racks and pipettors sold seperately. Note: The 25 and 50 ml tips require an adapter, which must be ordered separately.
Specifications
- Catalog No:
- 77898-452
- 77898-454
- 77898-450
- Environmentally Preferable:
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Environmental Certification:
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, Filter Tip Reloads reduce plastic waste by 33%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, Filter Tip Reloads reduce plastic waste by 33%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, Filter Tip Reloads reduce plastic waste by 33%