uTIP™ Sterile Filter Tip Reload Universal Compatible Pipette Tips
: Biotix
New Product
Biotix sterile uTIP™ filter tip reload universal pipette compatible tips with Flexfit™ lower insertion and ejection force by 70%.
uTIP™ filter tip reloads are manufactured in sustainable benchtop and user friendly packaging
- Reduce plastic waste by 30% and easily refill racks
- Fits universal pipettes
- Pure resin made of virgin polypropylene
- Certified free of RNase, DNase, Pyrogens, and PCR inhibitors
FlexFit™ technology features a flexible proximal end to ensure a secure seal with 66-80% less insertion force than competitor tips. Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip.
:
Tip refills only. Empty racks and pipettors sold seperately.
- Catalog No:
- 77898-444
- 77898-446
- 77898-442
- 77898-448
- 77898-438
- 77898-440
- 77898-456
- Environmentally Preferable:
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Waste Reducing, Sustainable Packaging
- Environmental Certification:
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, Filter Tip Reloads reduce plastic waste by 33%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, Filter Tip Reloads reduce plastic waste by 33%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, Filter Tip Reloads reduce plastic waste by 33%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, Filter Tip Reloads reduce plastic waste by 33%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, Filter Tip Reloads reduce plastic waste by 33%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, Filter Tip Reloads reduce plastic waste by 33%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, Filter Tip Reloads reduce plastic waste by 33%
Frequently Bought Together