ContactUsLinkComponent
Staphylococcus aureus (S. aureus) RN4220 Electroporation Competent Cells
Staphylococcus aureus (S. aureus) RN4220 Electroporation Competent Cells
  104090-478
undefined
Staphylococcus aureus (S. aureus) RN4220 Electroporation Competent Cells
  104090-478
 :  1294-40
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Size:
    5×200 µl
  • Storage Conditions:
    –80 °C
Intact Genomics (ig®) Staphylococcus aureus (S. aureus) RN4220 Electrocompetent Cells are optimized to provide high transformation efficiencies making them ideal for various applications.
  • Electrocompetent
  • Transformation efficiency >/= 1×10^5cfu/ug DNA
  • Intermediate cloning host
This strain is commonly used in studies involving virulence, resistance, metabolism and more.
Staphylococcus aureus RN4220 is characterized by a mutation in the sau1 hsdR genes, making it restriction deficient and hence an excellent intermediate cloning host.
 :
Manufactured under ISO 13485:2016 certified quality management system.