Intact Genomics TG1 Phage Display ElectroCompetent Cells are suitable for protein expression and preparation of antibody or peptide phage display libraries.
For antibody or peptide phage display libraries
- Transformation efficiency: ≥4×10¹⁰ cfu/µg pUC19 DNA
- Blue/white screening
Suitable for the production of phage display libraries.
Manufactured under ISO 13485:2016 certified quality management system.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 104090-460
- 104090-458
- Storage Conditions:
- –80 °C
- –80 °C