About this item
Highest transformation efficiencies.
Derivative of: DH5-alpha
- Transformation efficiency: ≥1×109 cfu/µg pUC19 DNA
- Blue/white screening: Yes
Intact Genomics 5 alpha chemically competent E. coli cells are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning. ig® 5 alpha chemically competent cells are at least twice the transformation efficiency of the nearest competitor.
Manufactured under ISO 13485:2016 certified quality management system.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 104090-440
- 104090-442
- Storage Conditions:
- –80 °C
- –80 °C