Enables development of more efficient plant transformation systems.
Optimized for the highest transformation efficiencies
- Requires supplemental methionine for growth
- Useful for transgenic operations of plants
- Octopine-type Ti plasmid pAL4404
Intact Genomics (ig®) Methionine-Auxotrophic LBA4404Met Agrobacterium chemically competent cells include modifications so that they will not grow unless free methionine is added to minimal medium.
This prevents the bacteria from overgrowing plant tissues when used for plant transformation.
Methionine- Auxotrophic LBA4404Met Agrobacterium chemically competent cells are optimized for the highest transformation efficiencies.
The LBA4404Met strain is useful for transgenic operations of tomatoes, tobacco and other plants.
LBA4404Met contains a rifampicin resistance gene (rif). LBA4404Met strain also contains an octopine-type Ti plasmid pAL4404Met without self-transport function, which contains the vir gene.
Manufactured under ISO 13485:2016 certified quality management system.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 104090-448
- 104090-446
- Storage Conditions:
- –80 °C
- –80 °C