The ig® RPA Kit version 1 from Intact Genomics gives the user a faster, more streamlined Recombinase Polymerase Amplification experience by providing the necessary reagents and enzymes to amplify user-provided DNA template. Isothermal RPA is an enhancement over PCR amplification because it does not require thermocycling and provides better sensitivity.
Highly selective and sensitive isothermal amplification technique
- Alternative to PCR
- No Thermocycler needed
- Speed and sensitivity, excellent for rapid molecular and agricultural tests
- No DNA pretreatment required
- Compatible with a variety of endpoint detection methods, (e.g. lateral flow, real-time fluorescence)
- All enzymes thoroughly tested for activity and purity
IG® Recombinase Polymerase Amplification (RPA) kit provides a mixture of the necessary enzymes to simplify and quicken the setup for your RPA reaction.
Manufactured under ISO 13485:2016 Quality Management certified standards.
Kit contains:
Enzyme mixture
2× RPA reaction master mix
280 mM Magnesium Acetate (MgOAc)
RPA positive control
Store at −20 °C.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- One step:Yes