Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant CXCL8 (IL-8) (from E. coli)
 
undefined
Human Recombinant CXCL8 (IL-8) (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Chemokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Avoid repeated freeze-thaw cycles 12 months from date of receipt, –20...–70 °C as supplied. 1 month, 2...8 °C under sterile conditions after reconstitution. 3 months, –20...–70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <0.01 EU per 1 μg of the protein by the LAL method
  • Biological Activity:
    EC50 = 0.083 nM determined by Migration Assay in cells expressing recombinant CXCR1
  • Gene ID:
    Accession # P10145 (28-99)
  • Reconstitution Instructions:
    Spin sample prior to reconstitution. Recommended concentration of 100 μg/ml in sterile water.
  • Endotoxin-free:
    Y
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Protein Synonyms:
    IL8|NAF|GCP1|LECT|LUCT|NAP1|GCP-1|LYNAP|MDNCF|MONAP|NAP-1|SCYB8
  • Protein/Peptide Name:
    CXCL8 (IL-8)
  • Purity:
    >97%
  • Molecular Weight:
    8.39 kDa
  • Sequence:
    SAKELRCQCIKTYSKPFHPKFIKELRVIESGPHCANTEIIVKLSDGRELCLDPKENWVQRVVEKFLKRAENS
  • Endotoxin Level:
    Very low
  • Formulation:
    Lyophilized pellet
  • Shipping Temperature:
    RT
  • Tested Applications:
    Migration assay

 

Frequently Bought Together