Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant CXCL12 (SDF-1a) (from E. coli), Biotinylated
 
undefined
Human Recombinant CXCL12 (SDF-1a) (from E. coli), Biotinylated
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Conjugation:
    Biotin
  • Protein Function:
    Chemokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Avoid repeated freeze-thaw cycles. 12 months from date of receipt, –20... –70 °C as supplied. ChemoTactics suggests using immediately after reconstitution. 1 month, –20...–70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <0.01 EU per 1 μg of the protein by the LAL method
  • Biological Activity:
    EC50 = 0.5 - 0.9 nM determined by Calcium Flux with human CXCR4 in U937 cells
  • Gene ID:
    Accession # P48061-2 (22-89)
  • Reconstitution Instructions:
    Spin sample prior to reconstitution. Recommended concentration of 100 μg/ml in sterile water.
  • Endotoxin-free:
    Y
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Protein Synonyms:
    IRH|PBSF|SDF1|TLSF|TPAR1|SCYB12
  • Protein/Peptide Name:
    CXCL12 (SDF-1a)
  • Purity:
    >97%
  • Molecular Weight:
    10.38 kDa
  • Endotoxin Level:
    Very low
  • Formulation:
    Lyophilized pellet
  • Shipping Temperature:
    RT
  • Tested Applications:
    Migration assay

 

Frequently Bought Together