Specifications
- Control Type:Analogue
- Description:AREC 7 magnetic hotplate stirrer
- Speed Range:100 - 1500 rpm
- W×D×H:160×270×100 mm
- Max. Stirring Capacity H₂O:15 L
- Plate Dimensions:180×180 mm
- Plate Material:Ceramic
- Power input/output:800 W
- Weight:3.5 kg
- Electrical:115 V/60 Hz
Specifications
About this item
Robust analog magnetic hotplate stirrer equipped with a 180 mm (7") ceramic plate, safe and reliable, designed to deliver excellent heating and stirring performance for basic laboratory applications.
Advanced heating technology ensures temperature homogeneity and stability from room temperature up to 550 °C
- Powerful brushless ensuring power and resistance over time
- Stirs up to 15 L volume (H₂O) from 100 up to 1500 rpm
- SpeedServo™ counter reaction technology maintains the speed constant as viscosity changes
- Robust ceramic top offers exceptional chemical and scratch resistance and it is easy to clean with a simple wipe-down
- A warning icon illuminates when the plate temperature exceeds 50 °C and the heating function is inactive
The AREC 7 ceramic hotplate stirrer is designed to deliver reliable performance for everyday laboratory applications, offering powerful heating and efficient stirring capabilities. Its advanced heating plate technology ensures uniform temperature control up to 550 °C, providing excellent heat stability for accurate sample preparation. Capable of stirring up to 15 L, the robust brushless motor operates smoothly across a wide speed range from 100 to 1500 rpm. Whether performing titrations or preparing samples and buffers, the SpeedServo™ counter-reaction technology maintains consistent stirring speeds even when the medium’s viscosity changes, ensuring reliable and repeatable results every time.
AREC 7 ceramic hotplate stirrer prioritises operator safety and durability. A clear warning icon illuminates when the plate temperature exceeds 50 °C and the heating function is inactive, minimising the risk of accidental burns. The robust structure is designed to withstand harsh laboratory environments and resist chemical corrosion. The ceramic plate is scratch-proof and simple to clean, ensuring long-lasting performance with minimal maintenance.
Housing material: Aluminium body, technopolymer enclosure
Stirring volume: 15 L
Stirring speed range: 100 to 1500 rpm
Motor: Brushless
Heating control: Analog
Torque compensation: SpeedServo™
Hot top warning system: Illuminated icon (when the temperature is above 50 °C)
CE marked.