ProBath® Water Bath
Catalog # 470357-538
Supplier: Revolutionary Science
New Product
Specifications
- Temperature range:Ambient to 90 °C
- Capacity:5.7 L
- Description:ProBath 150 water bath
- Exterior W×D×H:178×400×203 mm (7×15.75×8")
- Interior W×D×H:149×302×149 mm (5.875×11.875×5.875")
- Electrical:110 - 120 VAC, 50 - 60 Hz, 250 W, 2.0 A
Specifications
About this item
9½" vertical clearance easily accommodates tall items such as 1 L flasks or beakers without removing the cover.
Ideal for DNA extraction, CRISPR, and heat shock
- 300 series stainless steel chamber construction
- Simple 2-button operation
- Over and under temperature alarms
- Two year warranty
The ergonomic design has a small footprint to fit in tight bench spaces.
Delivery:
Supplied with an impact-resistant polycarbonate cover.