ContactUsLinkComponent
EVOM™ STX Calibration Solutions
EVOM™ STX Calibration Solutions
Catalog # 77664-328
New Product
CAS Number:  
undefined
EVOM™ STX Calibration Solutions
Catalog # 77664-328
Supplier Number:  EVOM-CAL
New Product
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    EVOM™ STX calibration solution, 125 ml
Specifications
About this item
Calibration solution for your TEER measuring system electrodes. This can be used for troubleshooting any issues you are having with electrodes.