Order Entry
United States
ContactUsLinkComponent
Human Recombinant Th17 Cell Polarizing Plus Protein Bundle (from E. coli)
Human Recombinant Th17 Cell Polarizing Plus Protein Bundle (from E. coli)
Catalog # 77753-614
Supplier:  VWR International
New Product
CAS Number:  
undefined
Human Recombinant Th17 Cell Polarizing Plus Protein Bundle (from E. coli)
Catalog # 77753-614
Supplier:  VWR International
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    At or below −20 °C
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein/Peptide Name:
    Th17
  • Endotoxin Level:
    Low
  • Formulation:
    NA
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

Specifications

About this item

Cytokines for polarizing human T cells into Th17 cells. T helper 17 cells (Th17) are a distinct type of T cell subset that are thought to be important in immunity against microbes at the epitilial and mucosal interfaces of the body.

  • High quality

  • Low endotoxin
  • Affordable

Disruption of the regular function or balance of Th17 cells is thought to be involved in several autoimmune diseases and/or opportunistic infections. Th17 cells are characterized by their ability to produce IL-17, IL-22 and IL-21. Th17 cells can be cultured from naive CD4+ T cells by culturing them in the presence of anti-CD3 and anti-CD28 stimulation with recombinant IL-6 and recombinant TGF-β 1. In some cases neutralizing antibodies against IL-12, IL-4 and IFNγ are used along with recombinat cytokines IL-1 β, IL-21 and/or IL-23.

Caution:

For research use only.