Order Entry
United States
ContactUsLinkComponent
Human Recombinant FLT-3 Ligand (from E. coli)
Human Recombinant FLT-3 Ligand (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant FLT-3 Ligand (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: OCI-Aml5 cell proliferation; ED₅₀ ≤5 ng/ml; Bioactivity ≥2.0×10⁵ units/mg
  • Gene ID:
    P49771
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Flt3L|Fms-related tyrosine kinase 3 ligand
  • Protein/Peptide Name:
    FLT-3 Ligand (AF)
  • Purity:
    ≥97%
  • Molecular Weight:
    17.7 kDa
  • Sequence:
    MTQDCSFQHS PISSDFAVKI RELSDYLLQD YPVTVASNLQ DEELCGGLWR LVLAQRWMER LKTVAGSKMQ GLLERVNTEI HFVTKCAFQP PPSCLRFVQT NISRLLQETS EQLVALKPWI TRQNFSRCLE LQCQPDSSTL PPPWSPRPLE ATAPT
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications

Frequently Bought Together