Rat Recombinant IL-6 (from E. coli)
New Product
Interleukin 6 (IL-6) is a cytokine that is expressed by T cells, macrophages, and muscle cells. IL-6 acts to stimulate an immune response upon infection or trauma. IL-6 has both pro-inflammatory and anti-inflammatory functions, is capable of promoting fever, and signals through the cell-surface type I cytokine receptor complex containing the IL-6Ra and gp130 chains.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Rat
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:Bioactivity assay: B9 cell proliferation; ED50 ≤2 ng/ml; Bioactivity ≥5.0×10⁵ units/mg
- Gene ID:P20607
- Reconstitution Instructions:Sterile 10 mM HCl at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:IFN-β2, B-cell Differentiation Factor, BSF-2, HSF, MGI-2
- Protein/Peptide Name:IL-6
- Purity:≥90%
- Molecular Weight:21.9 kDa
- Sequence:MFPTSQVRRG DFTEDTTHNR PVYTTSQVGG LITYVLREIL EMRKELCNGN SDCMNSDDAL SENNLKLPEI QRNDGCFQTG YNQEICLLKI CSGLLEFRFY LEFVKNNLQD NKKDKARVIQ SNTETLVHIF KQEIKDSYKI VLPTPTSNAL LMEKLESQKE WLRTKTIQLI LKALEEFLKV TMRSTRQT
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA) + 2:1 Mannitol to protein
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Frequently Bought Together