Mouse Recombinant IL-33 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 33 (IL-33) is a member of the IL-1 cytokine family and is constitutively expressed in smooth muscle and airway epithelial cells. IL-33 signals through the interleukin 1 receptor-like 1 (IL-1R1) and interleukin-1 receptor accessory protein (IL1RAP) receptors to ativate NF-κB and MAPK signaling pathways. IL-33 functions to induce type 2 cytokine production in polarized Type 2 helper T (Th2) cells.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:Bioactivity assay: D10S cell proliferation; ED50 ≤2 ng/ml; Bioactivity ≥5.0×10⁵ units/mg
- Gene ID:Q8BVZ5
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:NP-HEV
- Protein/Peptide Name:IL-33
- Purity:≥90%
- Molecular Weight:17.7 kDa
- Sequence:MSIQGTSLLT QSPASLSTYN DQSVSFVLEN GCYVINVDDS GKDQEQDQVL LRYYESPCPA SQSGDGVDGK KLMVNMSPIK DTDIWLHAND KDYSVELQRG DVSPPEQAFF VLHKKSSDFV SFECKNLPGT YIGVKDNQLA LVEEKDESCN NIMFKLSKI
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Frequently Bought Together