Mouse Recombinant IL-3 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells. IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: NFS-60 cell proliferation; ED50 ≤0.25 ng/ml; Bioactivity ≥4.0×10⁶ units/mg
- Gene ID:P01586
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:MCGF|Multi-CSF|HCGF|P-cell stimulating factor
- Protein/Peptide Name:IL-3
- Purity:≥95%
- Molecular Weight:15.2 kDa
- Sequence:MDTHRLTRTL NCSSIVKEII GKLPEPELKT DDEGPSLRNK SFRRVNLSKF VESQGEVDPE DRYVIKSNLQ KLNCCLPTSA NDSALPGVFI RDLDDFRKKL RFYMVHLNDL ETVLTSRPPQ PASGSVSPNR GTVEC
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Frequently Bought Together