Human Recombinant Fractalkine (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Fractalkine, also known as CX3CL1, is a cytokine protein containing a CX3C chemokine motif atop a mucin stalk. Fractalkine is produced by non-hemopoietic cells, including neurons and astrocytes. Soluble fractalkine functions as a chemoattractant for T cells and monocytes. Cell-membrane-bound fractalkine, which is induced on activated endothelial cells, promotes leukocyte adhesion. The transmembrane chemokine receptor CX3CR1 mediates the adhesive and chemoattractant functions of fractalkine.
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:−20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P78423
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:Neurotactin
- Protein/Peptide Name:Fractalkine
- Purity:≥95%
- Molecular Weight:8.6 kDa
- Sequence:#N/A
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Specifications
Frequently Bought Together