Order Entry
United States
ContactUsLinkComponent
Human Recombinant BDNF (from E. coli)
Human Recombinant BDNF (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant BDNF (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6); ED₅₀ ≤40 ng/ml; Bioactivity ≥2.5×10⁴ units/mg
  • Gene ID:
    P23560
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein/Peptide Name:
    BDNF
  • Purity:
    ≥95%
  • Molecular Weight:
    13.6/27.3 kDa
  • Sequence:
    MHSDPARRGE LSVCDSISEW VTAADKKTAV DMSGGTVTVL EKVPVSKGQL KQYFYETKCN PMGYTKEGCR GIDKRHWNSQ CRTTQSYVRA LTMDSKKRIG WRFIRIDTSC VCTLTIKRGR
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications

Frequently Bought Together