TubeWriter™ Care+ Barcode Upgrade
77871-082
New Product
- Description:TubeWriter™ Care+ barcode upgrade
An additional add-on to Care+ that enables 1D or 2D data matrix barcoding of samples.
The TubeWriter™ team will work with users to build a barcoding solution for sample labeling .
: Requires a TubeWriter™ Care+ subscription package.
: Upgrade includes: 1D barcodes, 2D data matrix barcodes, Custom images.