About this item
IL-10 is an anti-inflammatory cytokine produced by macrophages and type 2 T helper (Th2) cells. Inhibits the production of pro-inflammatory cytokines such as interferon gamma (IFNγ), tumor necrosis factor alpha (TNFα), interleukin 2 (IL-2), interleukin 3 (IL-3), interleukin 4 (IL-4), and granulocyte-macrophage colony-stimulating factor (GM-CSF), made by macrophages and regulatory T cells. Suppresses antigen presentation on antigen presenting cells, and enhances the survival, proliferation, and antibody production of B cells. The biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance.
- High quality Low endotoxin Lot-to-lot consistency Animal free Manufactured following GMP guidelines
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:At or below –20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.05
- Biological Activity:Bioactivity assay: Proliferation of MC/9 ; ED50 ≤5 ng/ml; Bioactivity ≥5.0×10³ IU/µg
- Gene ID:P22301
- Reconstitution Instructions:Sterile water at 0.1 - 0.5 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:B-TCGF|CSIF|TGIF
- Protein/Peptide Name:IL-10
- Purity:≥97%
- Molecular Weight:18.8
- Sequence:MSPGQGTQSE NSCTHFPGNL PNMLRDLRDA FSRVKTFFQM KDQLDNLLLK ESLLEDFKGY LGCQALSEMI QFYLEEVMPQ AENQDPDIKA HVNSLGENLK TLRLRLRRCH RFLPCENKSK AVEQVKNAFN KLQEKGIYKA MSEFDIFINY IEAYMTMKIR N
- Endotoxin Level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:GMP
- Shipping Temperature:Ambient