Human Recombinant PTN (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Pleiotrophin (PTN) is a heparin-binding growth factor that has mitogenic effects on fibroblast, epithelial, and endothelial cells. PTN is made by many tissues, but is predominantly secreted by nervous tissue during development. PTN induces neurite outgrowth and is involved in tumor growth and metastasis. PTN binds with low affinity to the cell surface receptor nucleolin to inhibit HIV-1 infection. PNT also binds the receptor protein tyrosine phosphatase type Z (PTPRZ), syndecan-3, and anaplastic lymphoma kinase (ALK) receptors.
- High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P21246
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:HBBM|HBGF-8|NEGF1|HARP
- Protein/Peptide Name:PTN
- Purity:≥95%
- Molecular Weight:15.4
- Sequence:MGKKEKPEKK VKKSDCGEWQ WSVCVPTSGD CGLGTREGTR TGAECKQTMK TQRCKIPCNW KKQFGAECKY QFQAWGECDL NTALKTRTGS LKRALHNAEC QKTVTISKPC GKLTKPKPQA ESKKKKKEGK KQEKMLD
- Endotoxin Level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Specifications
Frequently Bought Together