Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL-29 (from E. coli)
Human Recombinant IL-29 (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant IL-29 (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q8IU54
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    IFNλ1|Type III interferon|Interferon lambda|IFN-lambda
  • Protein/Peptide Name:
    IL-29
  • Purity:
    ≥95%
  • Molecular Weight:
    19.899999999999999
  • Sequence:
    MPTSKPTTTG KGCHIGRFKS LSPQELASFK KARDALEESL KLKNWSCSSP VFPGNWDLRL LQVRERPVAL EAELALTLKV LEAAAGPALE DVLDQPLHTL HHILSQLQAC IQPQPTAGPR PRGRLHHWLH RLQEAPKKES AGCLEASVTF NLFRLLTRDL KYVADGNLCL RTSTHPEST
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together