Mouse Recombinant IL-22 (from E. coli cells)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 22 (IL-22), also called IL-TIF, is an IL-10 family member that is produced by activated dendritic cells and T lymphocytes. IL-22 signals via a heteroduplex receptor consisting of IL-22R and IL-10RB chains. IL-22 is a potent mediator of cellular inflammatory responses and wound healing.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: Production of IL-10 from human COLO-205 cells; ED50 ≤1 ng/ml; Bioactivity ≥1.0×10⁶ units/mg
- Gene ID:Q9JJY9
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:IL-TIF
- Protein/Peptide Name:IL-22
- Purity:≥95%
- Molecular Weight:16.8 kDa
- Sequence:MLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACV
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Specifications
Frequently Bought Together