Mouse Recombinant IL-36 G (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 36 gamma (IL-36 ɣ) is a member of the interleukin 1 (IL-1) cytokine family and protects against pathogens in the skin, lung, and stomach epithelial barriers. IL-36 ɣ binds the interleukin-1 receptor accessory protein (IL-1RAcP) and the orphan IL-1R-related protein 2 (IL-1Rrp2) receptors to activate NF-kB and MAP kinase signaling pathways, resulting in the induced production of inflammatory cytokines and chemokines.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q8R460
- Reconstitution Instructions:Sterile 10 mM sterile HCl at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:IL-1F9|IL-1 epsilon
- Protein/Peptide Name:IL-36 G
- Purity:≥95%
- Molecular Weight:17.5 kDa
- Sequence:MGRETPDFGE VFDLDQQVWI FRNQALVTVP RSHRVTPVSV TILPCKYPES LEQDKGIAIY LGIQNPDKCL FCKEVNGHPT LLLKEEKILD LYHHPEPMKP FLFYHTRTGG TSTFESVAFP GHYIASSKTG NPIFLTSKKG EYYNINFNLD IKS
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Specifications
Frequently Bought Together