Human Recombinant FGF-6 (from E. coli)
About this item
Fibroblast growth factor 6 (FGF-6) is a heparin-binding growth factor that is expressed in epithelial and mesenchymal lineages. FGF-6 binds and signals through the FGF receptors FGFR1, FGFR2, and FGFR4. FGF-6 functions as a mitogen for vascular endothelial cells and fibroblasts. FGF-6 is also an important factor driving muscle differentiation and regeneration.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
5 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Fibroblast growth factor 6 (FGF-6) is a heparin-binding growth factor that is expressed in epithelial and mesenchymal lineages. FGF-6 binds and signals through the FGF receptors FGFR1, FGFR2, and FGFR4. FGF-6 functions as a mitogen for vascular endothelial cells and fibroblasts. FGF-6 is also an important factor driving muscle differentiation and regeneration.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Documents
VWR Cytokine and Growth Factors.pdf
Product Literature
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77751-014
- 77751-006
- 77751-008
- 77751-010
- 77751-012
- Protein/Peptide Name:
- FGF-6
- SDF-1 A/CXCL12
- FGF-6
- FGF-6
- FGF-6
- Protein Synonyms:
- Heparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6
- Heparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6
- Heparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6
- Heparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6
- Heparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P48061
- P10767
- P48061
- P48061
- P48061
- Sequence:
- MGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI
- MGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI
- MGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI
- MGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI
- MGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI
- Protease-free:
- N
- N
- N
- N
- N
- Animal-Free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-Free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin Level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin Content:
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5
- Molecular Weight:
- 18.9 kDa
- 18.9 kDa
- 18.9 kDa
- 18.9 kDa
- 18.9 kDa
- Reconstitution Instructions:
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Storage Conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping Temperature:
- RT
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77751-014Protein/Peptide NameFGF-6Protein SynonymsHeparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6Protein/Peptide TypeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP48061SequenceMGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRIProtease-freeNAnimal-FreeNNuclease-freeNCarrier-FreeYEndotoxin-freeNEndotoxin LevelLowEndotoxin ContentEndotoxin LAL (EU/µg) ≤0.1Biological ActivityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5Molecular Weight18.9 kDaReconstitution InstructionsSterile water at 0.1 mg/mlStorage Conditions–20 °CShipping TemperatureRT - Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77751-006Protein/Peptide NameSDF-1 A/CXCL12Protein SynonymsHeparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6Protein/Peptide TypeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP10767SequenceMGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRIProtease-freeNAnimal-FreeNNuclease-freeNCarrier-FreeYEndotoxin-freeNEndotoxin LevelLowEndotoxin ContentEndotoxin LAL (EU/µg) ≤0.1Biological ActivityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5Molecular Weight18.9 kDaReconstitution InstructionsSterile water at 0.1 mg/mlStorage Conditions–20 °CShipping TemperatureRT - Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77751-008Protein/Peptide NameFGF-6Protein SynonymsHeparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6Protein/Peptide TypeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP48061SequenceMGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRIProtease-freeNAnimal-FreeNNuclease-freeNCarrier-FreeYEndotoxin-freeNEndotoxin LevelLowEndotoxin ContentEndotoxin LAL (EU/µg) ≤0.1Biological ActivityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5Molecular Weight18.9 kDaReconstitution InstructionsSterile water at 0.1 mg/mlStorage Conditions–20 °CShipping TemperatureRT - Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77751-010Protein/Peptide NameFGF-6Protein SynonymsHeparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6Protein/Peptide TypeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP48061SequenceMGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRIProtease-freeNAnimal-FreeNNuclease-freeNCarrier-FreeYEndotoxin-freeNEndotoxin LevelLowEndotoxin ContentEndotoxin LAL (EU/µg) ≤0.1Biological ActivityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5Molecular Weight18.9 kDaReconstitution InstructionsSterile water at 0.1 mg/mlStorage Conditions–20 °CShipping TemperatureRT - Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77751-012Protein/Peptide NameFGF-6Protein SynonymsHeparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6Protein/Peptide TypeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP48061SequenceMGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRIProtease-freeNAnimal-FreeNNuclease-freeNCarrier-FreeYEndotoxin-freeNEndotoxin LevelLowEndotoxin ContentEndotoxin LAL (EU/µg) ≤0.1Biological ActivityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5Molecular Weight18.9 kDaReconstitution InstructionsSterile water at 0.1 mg/mlStorage Conditions–20 °CShipping TemperatureRT
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



