ContactUsLinkComponent
NexION® 5000 Multi-Quadrupole ICP Mass Spectrometer
NexION® 5000 Multi-Quadrupole ICP Mass Spectrometer
Catalog # 77865-774
Supplier:  PerkinElmer
New Product
undefined
NexION® 5000 Multi-Quadrupole ICP Mass Spectrometer
Catalog # 77865-774
Supplier:  PerkinElmer
Supplier Number:  N8160009
New Product
Specifications
  • Description:
    Spectrometer NexION® 5000 ICP-MS
Specifications
About this item
The first in its category to boast four quads, the NexION® 5000 multi-quadrupole ICP-MS is innovatively designed to meet and exceed the demanding requirements of ultra-trace elemental applications.
  • Innovatively designed
It delivers exceptionally low background equivalent concentrations and outstanding detection limits, key to ensuring accurate, repeatable results. Plus, it provides superior interference removal, phenomenal stability and unmatched matrix tolerance, for results you can count on. Discover how the NexION® 5000 takes ICP-MS performance beyond traditional triple quad.
Weight: 191.0 kg