ContactUsLinkComponent
PinAAcle™ 500 Flame Atomic Absorption Spectrometer
PinAAcle™ 500 Flame Atomic Absorption Spectrometer
Catalog # 77865-770
Supplier:  PerkinElmer
New Product
undefined
PinAAcle™ 500 Flame Atomic Absorption Spectrometer
Catalog # 77865-770
Supplier:  PerkinElmer
Supplier Number:  N3180011
New Product
Specifications
  • Description:
    Spectrometer PinAAcle™ 500 Touch AA
Specifications
About this item
Our PinAAcle™ 500 is the world’s first completely corrosion-resistant flame atomic absorption (AA) spectrometer, designed to withstand the harshest environments and most corrosive samples. It offers superior durability, longer life, lower maintenance costs, and the fastest return on investment of any flame AA.
  • Superior durability
A robust, fully-integrated, flame-only benchtop system, ideal for laboratories needing a reliable, easy-to-use, yet high-performance flame AA. It is equipped with an innovative touchscreen interface with the flexibility to operate via its easy-to-use Syngistix Touch™ software or the more comprehensive, optional Syngistix™ for AA Software.
Weight: 57.0 kg