Human Recombinant WISP-2 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
WNT1-inducible-signaling pathway protein 2 (WISP-2) is a member of the CYR61/CTGF/NOV (CCN) family of regulatory factors. WISP-2 is expressed in ectodermal, mesodermal, and endodermal lineages, including primary osteoblasts, fibroblasts, mesenchymal stem cells, and adipogenic precursor cells. WISP-2 is a canonical WNT ligand that regulates cell proliferation, adhesion, and metastasis. Secreted WISP-2 promotes mesenchymal precursor cell proliferation and maintains them in an undifferentiated state. In bone-forming osteoblasts, WISP-2 promotes osteoblast adhesion and inhibits osteocalcin production.
High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:O76076
- Reconstitution Instructions:Sterile 10 mM acetic acid at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:CTGF-L|Connective tissue growth factor-like protein|CCN5|CT58
- Protein/Peptide Name:WISP-2
- Purity:≥95%
- Molecular Weight:24.4 kDa
- Sequence:MQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAF
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Specifications
Frequently Bought Together