Mouse Recombinant IL-17AF (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 17AF (IL-17AF) is a heterodimer that is composed of the interleukin 17A (IL-17A) and interleukin 17F (IL-17F) members of the IL-17 family of cytokines. IL-17AF is produced by T helper 17 cells (Th17) following interleukin 23 (IL-23) stimulation. IL-17AF signals through the IL-17RA/IL-17RC receptor complex and functions to regulate inflammatory responses. IL-17AF induces chemokine and airway neutrophilia production, similar in function to IL-17A and IL-17F homodimers. In regard to these functions, IL-17AF is less active than the IL-17A homodimer and shows greater activity than the IL-17F homodimer. Human and rat IL-17AF are active on mouse cells.
High quality
Low endotoxin
Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q62386/Q7TNI7
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein/Peptide Name:IL-17AF
- Purity:≥95%
- Molecular Weight:30.1 kDa
- Sequence:IL-17A: MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA IL-17F: MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAA
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Specifications
Frequently Bought Together