Mouse Recombinant IGF-I (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Insulin-like growth factor I (IGF-I) is a growth factor that is produced by the liver. IGF-1 production is stimulated by growth hormone (GH). IGF-I binds the insulin-like growth factor 1 receptor (IGF1R) and the insulin receptor to stimulate systemic body growth. IGF-I is one of the most potent activators of the AKT signaling pathway, which stimulates cell proliferation and inhibits programmed cell death.
High quality
Low endotoxin
Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: FDC-P1 cell proliferation; ED50 ≤20 ng/ml; Bioactivity ≥5.0×10⁴ units/mg
- Gene ID:Q8CAR0
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:Somatomedin C|mechano growth factor|IGF-IA
- Protein/Peptide Name:IGF-I
- Purity:≥95%
- Molecular Weight:7.7 kDa
- Sequence:GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKAA
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Frequently Bought Together