Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL-31 (from E. coli)
Human Recombinant IL-31 (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant IL-31 (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: STAT3 Activation in U-87 MG cells; ED50 ≤Detectable at ≤25 ng/ml; Bioactivity ≥0
  • Gene ID:
    Q6EBC2
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein/Peptide Name:
    IL-31
  • Purity:
    ≥95%
  • Molecular Weight:
    15.8 kDa
  • Sequence:
    SHTLPVRLLR PSDDVQKIVE ELQSLSKMLL KDVEEEKGVL VSQNYTLPCL SPDAQPPNNI HSPAIRAYLK TIRQLDNKSV IDEIIEHLDK LIFQDAPETN ISVPTDTHEC KRFILTISQQ FSECMDLALK SLTSGAQQAT T
  • Endotoxin Level:
    Low
  • Formulation:
    20 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications

Frequently Bought Together