Human Recombinant TRAIL (from E. coli)
New Product
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a member of the tumor necrosis factor (TNF) family of cytokines. TRAIL is widely produced by a variety of cell types including tumor cells, smooth muscle of the lung and spleen, cerebellar glial cells, and thyroid follicular cells. TRAIL is a cytotoxic protein that induces apoptosis in tumor cells through the activation of the death receptors DR4 and DR5. TRAIL also binds the neutralizing decoy receptors, DcR1 and DcR2. Human TRAIL is active on mouse cells.
- High quality Low endotoxin Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P50591
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:Apo-2L|TL2|Apo-2 Ligand|TNFSF10|CD253
- Protein/Peptide Name:TRAIL
- Purity:≥90%
- Molecular Weight:19.5 kDa
- Sequence:MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Frequently Bought Together