ContactUsLinkComponent
Brady® SPC® Absorbents Acid Neutralizing Polymer
Brady® SPC® Absorbents Acid Neutralizing Polymer
Catalog # 77775-244
Supplier:  Brady Worldwide
New Product
Brady® SPC® Absorbents Acid Neutralizing Polymer
Catalog # 77775-244
Supplier:  Brady Worldwide
Supplier Number:  123064
New Product
Specifications
  • Description:
    Acid neutralizing polymer
Specifications
About this item
The acid neutralizer is used to solidify a wide range of base spills.
  • Class: chemical
  • Container type: bottle
The acid neutralizer povides safe spill control for most acids, including battery spills. Polymers neutralize and change color to let users know neutralization has taken place. These two pound shakers are non-toxic, non-corrosive, and non-hazardous. Acid neutralizer turns purple to indicate neutralization is complete.
Minimum order quantitiy: 10.