Specifications
- Color:Orange, Black
- Description:Centurion™ Hi-Vis reinforced flood barriers
- Diameter ×L:6"×10'
Specifications
About this item
Transform flood readiness and maximize asset protection. Enjoy rapid response and none of the labor or mess of sandbags with water-activated Centurion™ Hi-Vis Reinforced Flood Barriers.
Class: fresh water
- Polypropylene fill material
- Lint-free material texture
- Absorption capacity: 80 gal.
Centurion™ freshwater-swelled barriers contain, control, and divert flood water. Absorbent-filled design swells to full size on contact with water in 10 minutes. Flat and lightweight when dry for space-saving storage and easy positioning before first use. Easier and cleaner than sandbags. Built-in wedge prevents rolling . To stack barriers, place wedge on top of wedge with two barriers, then place a third barrier on top in the middle. Cannot be used with salt water. Hi-vis, heavy-duty orange mesh provides greater visibility and protects barrier from damage caused by dragging or debris.