Abgene™ Seals and Sealing Tape
New Product
Thermo Scientific™ Abgene™ seals minimize evaporation and protect samples from contamination and spilling. Adhesive seals seal all microplate formats and are ideal for all assay applications. Heat seals for ALPS™ microplate heat sealing instruments are ideal for storage applications, compound management, and high-throughput screenings including PCR.
Adhesive and heat seals
- Temperature range −80 to +120 °C
- Clear, white, and silver
- Pierceable options
- Peelable options
- Gas permeable options
- Heat seals designed for ALPS™ microplate heat sealers and generally compatible with other microplate heat sealers
Thermo Scientific™ Abgene™ heat seals provide an extra level of security over adhesive seals and cap mats due to a more uniform sealing. Plates need a raised rim around each well to enable the bonding of the seal to the well.
Frequently Bought Together