ContactUsLinkComponent
5-Methoxy-Uridine-5-Triphosphate (5-Methoxy-UTP)
5-Methoxy-Uridine-5-Triphosphate (5-Methoxy-UTP)
Catalog # 77868-094
New Product
CAS Number:  
undefined
5-Methoxy-Uridine-5-Triphosphate (5-Methoxy-UTP)
Catalog # 77868-094
Supplier Number:  N0434S
New Product
CAS Number:  
Specifications
  • Description:
    5-Methoxy-Uridine-5-Triphosphate (5-Methoxy-UTP), 100 mm concentration
  • Volume:
    0.1 ml
Specifications
About this item
mRNA has emerged as a potent modality for therapeutics.
  • Supplied as sodium salts, pH 7.0
  • Modified NTPs are commonly used for reduction of immunogenicity for in vitro transcription RNA
  • No detectable RNase, DNase, phosphatase or protease contaminating activities
  • ≥95% purity, as determined by HPLC
  • Optimal activity and stability for up to 24 months
Incorporation of chemically modified ribonucleotides improves the efficacy of these therapies by increasing stability, reducing immunogenicity, and enhancing translatability of mRNAs.