Order Entry
United States
ContactUsLinkComponent
Pseudouridine-5-Triphosphate (Pseudo-UTP)
Pseudouridine-5-Triphosphate (Pseudo-UTP)
Catalog # 77868-092
New Product
CAS Number:  
undefined
Pseudouridine-5-Triphosphate (Pseudo-UTP)
Catalog # 77868-092
Supplier Number:  N0433S
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Pseudouridine-5-Triphosphate (Pseudo-UTP), 100 mm concentration
  • Volume:
    0.1 ml

Specifications

About this item

mRNA has emerged as a potent modality for therapeutics.

  • Supplied as sodium salts, pH 7.0

  • Modified NTPs are commonly used for reduction of immunogenicity for in vitro transcription RNA
  • No detectable RNase, DNase, phosphatase or protease contaminating activities
  • ≥95% purity, as determined by HPLC
  • Optimal activity and stability for up to 24 months

Incorporation of chemically modified ribonucleotides improves the efficacy of these therapies by increasing stability, reducing immunogenicity, and enhancing translatability of mRNAs.