Anti-Human Metapneumovirus Recombinant Monoclonal Antibody [clone: RV47]
New Product
This antibody has been tested to bind to fusion (F) glycoprotein of human Metapneumovirus (hMPV) subtype A1 strain (Cat. No. Q82X76-A) or B1 strain (Cat. No. Q82X76-B).
Type: Primary
Antigen: hMPV
Clonality: Recombinant monoclonal
Clone: RV47
Conjugation:
Epitope:
Host: Human
Isotype: IgA1 Kappa
Reactivity: Human
- Catalog No:
- 77761-640
- 77761-642
- Antigen Symbol:
- hMPV
- hMPV
- Antigen Name:
- human Metapneumovirus
- human Metapneumovirus
- Antigen Synonyms:
- Fusion (F) glycoprotein
- Fusion (F) glycoprotein
- Antibody Type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV47
- RV47
- Reactivity:
- Human
- Human
- Host:
- Human
- Human
- Isotype:
- IgA1 kappa
- IgA1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Concentration:
- 1 mg/ml by refractometry
- 1 mg/ml by refractometry
- Storage Buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage Temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping Temperature:
- Ice
- Ice
Frequently Bought Together