Anti-Marburg Virus Human Recombinant Monoclonal Antibody [clone: RV53]
Supplier: ReVacc Scientific
New Product
Certificates
About this item
This antibody has been tested to bind to Marburg Virus Glycoprotein GP (Catalog # I27A51) and neutralize against Marburg Pseudovirus (Cat. No. N31O81) on Vero cells.
Type: Primary
Antigen: MARV
Clonality: Recombinant monoclonal
Clone: RV53
Conjugation:
Epitope:
Host: Human
Isotype: IgG1 Lambda
Reactivity: Human
Specifications
- Catalog No:
- 77761-666
- 77761-668
- Antigen Symbol:
- MARV
- MARV
- Antigen Name:
- Marburg Virus
- Marburg Virus
- Antigen Synonyms:
- Glycoprotein (GP)
- Glycoprotein (GP)
- Antibody Type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV53
- RV53
- Reactivity:
- Human
- Human
- Host:
- Human
- Human
- Isotype:
- IgG1 lambda
- IgG1 lambda
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Molecular Weight:
- 150 kDa
- 150 kDa
- Purification:
- Purified by Protein A chromatography
- Purified by Protein A chromatography
- Concentration:
- 1 mg/ml by refractometry
- 1 mg/ml by refractometry
- Storage Buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage Temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping Temperature:
- Ice
- Ice
Specifications
Frequently Bought Together